accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2216-84-4

IUPAC Name: cyclohexane-1,2,3,4,5,6-hexacarboxylic acid
Synonyms: cyclohexane-1,2,3,4,5,6-hexacarboxylic acid
Molecular Weight (g/mol): 348.22
Molecular Formula: C12H12O12
InChi Key: DTGRIEIJTWNZQF-UHFFFAOYSA-N
SMILES: OC(=O)C1C(C(C(C(C1C(O)=O)C(O)=O)C(O)=O)C(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results