accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2217-02-9

IUPAC Name: (1S,2S,4R)-1,3,3-trimethylbicyclo[2.2.1]heptan-2-ol
Synonyms: fenchyl alcohol ((endo)-(+-))-fenchol (endo)-fenchol (-)-endo-fenchol endo-fenchol α-fenchol 2-fenchanol (exo)-fenchol ((exo)-(+-))-fenchol α-fenchyl alcohol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: IAIHUHQCLTYTSF-MRTMQBJTSA-N
SMILES: CC1(C)[C@@H]2CC[C@@](C)(C2)[C@@H]1O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Concentration 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)