accessibility menu, dialog, popup

UserName

Viewing profile as user:
II

CAS RN 2217-79-0

IUPAC Name: diphenyliodanium iodide
Synonyms: diphenyliodanium iodide
Molecular Weight (g/mol): 408.02
Molecular Formula: C12H10I2
InChi Key: WQIRVUAXANLUPO-UHFFFAOYSA-M
SMILES: [I-].[I+](C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results