accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2218-80-6

CuOOOO

CAS RN 2218-80-6

IUPAC Name: copper(2+) bis(4-cyclohexylbutanoate)
Synonyms: copper(2+) bis(4-cyclohexylbutanoate)
Molecular Weight (g/mol): 402.03
Molecular Formula: C20H34CuO4
InChi Key: FGUOYHAMMRMUQO-UHFFFAOYSA-L
SMILES: [Cu++].[O-]C(=O)CCCC1CCCCC1.[O-]C(=O)CCCC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results