accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22180-53-6

IUPAC Name: palladium(2+) bis(triphenylphosphane) dibromide
Synonyms: palladium(2+) bis(triphenylphosphine) dibromide
Molecular Weight (g/mol): 790.81
Molecular Formula: C36H30Br2P2Pd
InChi Key: MCSDDEAMPOYJJI-UHFFFAOYSA-L
SMILES: [Br-].[Br-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results