accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 221884-63-5

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pent-4-enoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pent-4-enoic acid
Molecular Weight (g/mol): 337.38
Molecular Formula: C20H19NO4
InChi Key: YVBLQCANYSFEBN-UHFFFAOYNA-N
SMILES: OC(=O)C(CC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)