accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2219-82-1

CH3HOH3CH3CCH3

CAS RN 2219-82-1

IUPAC Name: 2-tert-butyl-6-methylphenol
Synonyms: 2-tert-butyl-6-methylphenol
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: BKZXZGWHTRCFPX-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1O)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results