accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22195-47-7

IUPAC Name: 1-(2,2-dimethyl-1,3-dioxolan-4-yl)methanamine
Synonyms: 1-(2,2-dimethyl-1,3-dioxolan-4-yl)methanamine
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: HXOYWCSTHVTLOW-UHFFFAOYNA-N
SMILES: CC1(C)OCC(CN)O1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)