accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22201-45-2

IUPAC Name: 4-tert-butylbenzoyl 4-tert-butylbenzoate
Synonyms: 4-tert-butylbenzoyl 4-tert-butylbenzoate
Molecular Weight (g/mol): 338.45
Molecular Formula: C22H26O3
InChi Key: KKDPRVYYZZUPLR-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C(=O)OC(=O)C1=CC=C(C=C1)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results