accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOOOOO

CAS RN 22202-68-2

IUPAC Name: disodium 2-oxopentanedioate
Synonyms: disodium α ketoglutarate
Molecular Weight (g/mol): 190.06
Molecular Formula: C5H4Na2O5
InChi Key: YBGBJYVHJTVUSL-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)CCC(=O)C([O-])=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results