accessibility menu, dialog, popup

UserName

Viewing profile as user:
CuCuS

CAS RN 22205-45-4

IUPAC Name: bis(λ¹-copper(1+)) sulfanediide
Synonyms: copper(I) sulfide|cuprous sulfide|copper(1+) sulfide|dicopper sulphide|kupfer(I)-sulfid|cuprasulfide|dicopper monosulfide
Molecular Weight (g/mol): 159.15
Molecular Formula: Cu2S
InChi Key: AQMRBJNRFUQADD-UHFFFAOYSA-N
SMILES: [S--].[Cu+].[Cu+]
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)