accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3H3CH3CH2OHF

CAS RN 22206-57-1

IUPAC Name: tetrabutylazanium hydrate hydrofluoride
Synonyms: tetrabutylammonium ion hydrate hydrofluoride
Molecular Weight (g/mol): 280.49
Molecular Formula: C16H39FNO
InChi Key: UQCWXKSHRQJGPH-UHFFFAOYSA-N
SMILES: O.F.CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)