CAS RN 2221-13-8
IUPAC Name:
3-phenylimidazolidine-2,4-dione
Synonyms:
N-phenyl-2,5-imidazolidione
Molecular Weight (g/mol):
176.18
Molecular Formula:
C9H8N2O2
InChi Key:
RUEGAVIENIPHGK-UHFFFAOYSA-N
SMILES:
O=C1CNC(=O)N1C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
3-Phenylhydantoin, 95%
3-Phenylhydantoin, 95%
CAS: 2221-13-8 Molecular Formula: C9H8N2O2 Molecular Weight (g/mol): 176.18 MDL Number: MFCD00209486 InChI Key: RUEGAVIENIPHGK-UHFFFAOYSA-N PubChem CID: 227930 IUPAC Name: 3-phenylimidazolidine-2,4-dione SMILES: O=C1CNC(=O)N1C1=CC=CC=C1