accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 222180-19-0

OOHO

CAS RN 222180-19-0

IUPAC Name: 3'-formyl-[1,1'-biphenyl]-3-carboxylic acid
Synonyms: 3'-formyl-[1,1'-biphenyl]-3-carboxylic acid
Molecular Weight (g/mol): 226.23
Molecular Formula: C14H10O3
InChi Key: QOCUSJPHQSKJJR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1)C1=CC(C=O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results