accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3C

CAS RN 222319-05-3

IUPAC Name: 9,9-dimethyl-N2,N7-bis(naphthalen-1-yl)-N2,N7-diphenyl-9H-fluorene-2,7-diamine
Synonyms: 9,9-dimethyl-N2,N7-bis(naphthalen-1-yl)-N2,N7-diphenylfluorene-2,7-diamine
Molecular Weight (g/mol): 628.82
Molecular Formula: C47H36N2
InChi Key: KJEQVQJWXVHKGT-UHFFFAOYSA-N
SMILES: CC1(C)C2=C(C=CC(=C2)N(C2=CC=CC=C2)C2=C3C=CC=CC3=CC=C2)C2=C1C=C(C=C2)N(C1=CC=CC=C1)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results