accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOONSH3CH3CO

CAS RN 22236-45-9

IUPAC Name: dimethyl[(4-methylbenzenesulfonyl)imino]-λ⁶-sulfanone
Synonyms: dimethyl[(4-methylbenzenesulfonyl)imino]-λ⁶-sulfanone
Molecular Weight (g/mol): 247.33
Molecular Formula: C9H13NO3S2
InChi Key: IRNAWARRPQUZDU-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N=S(C)(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results