accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22246-18-0

IUPAC Name: 7-hydroxy-1,2,3,4-tetrahydroquinolin-2-one
Synonyms: 7-hydroxy-3,4-dihydro-1H-quinolin-2-one
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: LKLSFDWYIBUGNT-UHFFFAOYSA-N
SMILES: OC1=CC2=C(CCC(=O)N2)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)