accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2225-23-2

IUPAC Name: 1-methyl-4-[({2-[(4-methylbenzenesulfonyl)sulfanyl]ethyl}sulfanyl)sulfonyl]benzene
Synonyms: 1-methyl-4-({2-[(4-methylbenzenesulfonyl)sulfanyl]ethyl}sulfanylsulfonyl)benzene
Molecular Weight (g/mol): 402.56
Molecular Formula: C16H18O4S4
InChi Key: DUFUGAKEFZRFEQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)SCCSS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results