accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22254-24-6

IUPAC Name: (1R,5S)-3-[(3-hydroxy-2-phenylpropanoyl)oxy]-8-methyl-8-(propan-2-yl)-8-azabicyclo[3.2.1]octan-8-ium bromide
Synonyms: ipratropium bromide
Molecular Weight (g/mol): 412.37
Molecular Formula: C20H30BrNO3
InChi Key: LHLMOSXCXGLMMN-CLTUNHJMSA-M
SMILES: [Br-].CC(C)[N+]1(C)[C@H]2CC[C@@H]1CC(C2)OC(=O)C(CO)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results