accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3OCH3(E/Z)

CAS RN 22255-22-7

IUPAC Name: 1,3-dimethoxy-5-[2-(4-methoxyphenyl)ethenyl]benzene
Synonyms: 1,3-dimethoxy-5-[2-(4-methoxyphenyl)ethenyl]benzene
Molecular Weight (g/mol): 270.33
Molecular Formula: C17H18O3
InChi Key: GDHNBPHYVRHYCC-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=CC2=CC(OC)=CC(OC)=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results