accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH4NH4OOOO

CAS RN 2226-88-2

IUPAC Name: diammonium butanedioate
Synonyms: diammonium succinate
Molecular Weight (g/mol): 152.15
Molecular Formula: C4H12N2O4
InChi Key: NHJPVZLSLOHJDM-UHFFFAOYSA-N
SMILES: [NH4+].[NH4+].[O-]C(=O)CCC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)