accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBrBr

CAS RN 22262-60-8

IUPAC Name: methyl 3-bromo-2-(bromomethyl)propanoate
Synonyms: methyl 3-bromo-2-(bromomethyl)propanoate
Molecular Weight (g/mol): 259.93
Molecular Formula: C5H8Br2O2
InChi Key: USXVPPOARMSYGY-UHFFFAOYSA-N
SMILES: COC(=O)C(CBr)CBr
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)