accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NClNOO

CAS RN 22280-56-4

IUPAC Name: 2-chloro-3-methyl-5-nitropyridine
Synonyms: 2-chloro-3-methyl-5-nitropyridine
Molecular Weight (g/mol): 172.57
Molecular Formula: C6H5ClN2O2
InChi Key: OSIOIGXJUZTWRI-UHFFFAOYSA-N
SMILES: CC1=CC(=CN=C1Cl)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)