CAS RN 22282-69-5
IUPAC Name:
2-fluoro-1,4-dihydropyridin-4-one
Synonyms:
2-fluoro-1H-pyridin-4-one
Molecular Weight (g/mol):
113.09
Molecular Formula:
C5H4FNO
InChi Key:
BRLSDLHMGRDAPF-UHFFFAOYSA-N
SMILES:
FC1=CC(=O)C=CN1
1
–
1
of
1
results
1
Promotions Available
2-Fluoro-4-hydroxypyridine, 97%
2-Fluoro-4-hydroxypyridine, 97%
CAS: 22282-69-5 Molecular Formula: C5H4FNO Molecular Weight (g/mol): 113.091 MDL Number: MFCD04114159 InChI Key: BRLSDLHMGRDAPF-UHFFFAOYSA-N Synonym: 2-fluoropyridin-4-ol,2-fluoro-4-hydroxypyridine,2-fluoro-pyridin-4-ol,4-pyridinol, 2-fluoro,2-fluoro-4-pyridinol,4 1h-pyridinone,2-fluoro,2-fluoranyl-1h-pyridin-4-one,4 1h-pyridinone, 2-fluoro,4 1h-pyridinone, 2-fluoro-9ci PubChem CID: 2762834 IUPAC Name: 2-fluoro-1H-pyridin-4-one SMILES: C1=CNC(=CC1=O)F