accessibility menu, dialog, popup

UserName

Viewing profile as user:
FIN

CAS RN 22282-70-8

IUPAC Name: 2-fluoro-4-iodopyridine
Synonyms: 2-fluoro-4-iodopyridine
Molecular Weight (g/mol): 222.99
Molecular Formula: C5H3FIN
InChi Key: ADPRIAVYIGHFSO-UHFFFAOYSA-N
SMILES: FC1=CC(I)=CC=N1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)