accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NBrNOO

CAS RN 22282-96-8

IUPAC Name: 6-bromo-2-methyl-3-nitropyridine
Synonyms: 6-bromo-2-methyl-3-nitropyridine
Molecular Weight (g/mol): 217.02
Molecular Formula: C6H5BrN2O2
InChi Key: PCNVKBBKROTUNS-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(Br)=N1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results