accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrBrCH3NCH3NOONNH3CH3COOHH

CAS RN 22291-04-9

IUPAC Name: dihydrogen 6,13-bis[2-(dimethylamino)ethyl]-6,13-diazatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1,3,8,10,15-pentaene-5,7,12,14-tetrone dibromide
Synonyms: dihydrogen 6,13-bis[2-(dimethylamino)ethyl]-6,13-diazatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1,3,8,10,15-pentaene-5,7,12,14-tetrone dibromide
Molecular Weight (g/mol): 570.28
Molecular Formula: C22H26Br2N4O4
InChi Key: URQFRTBLIAJQSB-UHFFFAOYSA-N
SMILES: [H+].[H+].[Br-].[Br-].CN(C)CCN1C(=O)C2=CC=C3C(=O)N(CCN(C)C)C(=O)C4=CC=C(C1=O)C2=C34
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results