accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNOO

CAS RN 22300-56-7

IUPAC Name: 5-methyl-2-phenyl-2H-1,2,3-triazole-4-carboxylate
Synonyms: 5-methyl-2-phenyl-1,2,3-triazole-4-carboxylate
Molecular Weight (g/mol): 202.19
Molecular Formula: C10H8N3O2
InChi Key: GQYQHAGPALBYDO-UHFFFAOYSA-M
SMILES: CC1=NN(N=C1C([O-])=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results