CAS RN 2230788-58-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sigma Aldrich [(1,3,5,7-Tetramethyl-6-Phenyl-2,4,6-Trioxa-6-Phosphaadamantane)-2-(2'-Amino-1,1'-Biphenyl)]Palladium(II) Methanesulfonate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.