accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 223127-47-7

IUPAC Name: 4-[5-(trifluoromethyl)pyridin-2-yl]benzoic acid
Synonyms: 4-[5-(trifluoromethyl)pyridin-2-yl]benzoic acid
Molecular Weight (g/mol): 267.21
Molecular Formula: C13H8F3NO2
InChi Key: FSLUYPHZCSKJJZ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=NC=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results