accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOCl

CAS RN 223128-33-4

IUPAC Name: 2-[4-(propan-2-yl)phenoxy]acetyl chloride
Synonyms: 2-(4-isopropylphenoxy)acetyl chloride
Molecular Weight (g/mol): 212.67
Molecular Formula: C11H13ClO2
InChi Key: YGVLHWFQKYVLFY-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(OCC(Cl)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results