accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OOHOHBa

CAS RN 22326-55-2

IUPAC Name: barium(2+) hydrate dihydroxide
Synonyms: barium(2+) hydrate dihydroxide
Molecular Weight (g/mol): 189.36
Molecular Formula: BaH4O3
InChi Key: GKQTUHKAQKWLIN-UHFFFAOYSA-L
SMILES: O.[OH-].[OH-].[Ba++]
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)