accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3CHO

CAS RN 2233-18-3

IUPAC Name: 4-hydroxy-3,5-dimethylbenzaldehyde
Synonyms: 4-hydroxy-3,5-dimethylbenzaldehyde
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: UYGBSRJODQHNLQ-UHFFFAOYSA-N
SMILES: CC1=CC(C=O)=CC(C)=C1O
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)