accessibility menu, dialog, popup

UserName

Viewing profile as user:
ZrH3CCH3OOH3CCH3OOH3CCH3OOH3CCH3OO

CAS RN 2233-42-3

IUPAC Name: zirconium(4+) tetrakis(2-ethylhexanoate)
Synonyms: zirconium(4+) tetrakis(2-ethylhexanoate)
Molecular Weight (g/mol): 664.05
Molecular Formula: C32H60O8Zr
InChi Key: OFYFURKXMHQOGG-UHFFFAOYNA-J
SMILES: [Zr+4].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results