accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2234-20-0

IUPAC Name: 1-ethenyl-2,4-dimethylbenzene
Synonyms: styrene, 2,4-dimethyl-
Molecular Weight (g/mol): 132.21
Molecular Formula: C10H12
InChi Key: OEVVKKAVYQFQNV-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C)C(C)=C1
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)