accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 223437-05-6

IUPAC Name: 1-methyl-1-propylpyrrolidin-1-ium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: 1-methyl-1-propylpyrrolidin-1-ium; bistriflylimide anion
Molecular Weight (g/mol): 408.37
Molecular Formula: C10H18F6N2O4S2
InChi Key: DKNRELLLVOYIIB-UHFFFAOYSA-N
SMILES: CCC[N+]1(C)CCCC1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results