accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOBOBOFFFFFFFFF

CAS RN 223440-94-6

IUPAC Name: tris(3,4,5-trifluorophenyl)-1,3,5,2,4,6-trioxatriborinane
Synonyms: tris(3,4,5-trifluorophenyl)-1,3,5,2,4,6-trioxatriborinane
Molecular Weight (g/mol): 473.66
Molecular Formula: C18H6B3F9O3
InChi Key: SXINGRFBONUWIF-UHFFFAOYSA-N
SMILES: FC1=CC(=CC(F)=C1F)B1OB(OB(O1)C1=CC(F)=C(F)C(F)=C1)C1=CC(F)=C(F)C(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results