accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrIN

CAS RN 223463-13-6

IUPAC Name: 5-bromo-2-iodopyridine
Synonyms: 5-bromo-2-iodopyridine
Molecular Weight (g/mol): 283.89
Molecular Formula: C5H3BrIN
InChi Key: HSNBRDZXJMPDGH-UHFFFAOYSA-N
SMILES: BrC1=CC=C(I)N=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (4)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (4)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (3)
  • (4)
  • (2)
  • (1)
  • (4)
  • (1)
  • (3)
  • (3)
  • (4)
  • (2)