accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 223463-14-7

IUPAC Name: (6-bromopyridin-3-yl)boronic acid
Synonyms: 6-bromopyridin-3-ylboronic acid
Molecular Weight (g/mol): 201.81
Molecular Formula: C5H5BBrNO2
InChi Key: BCYWDUVHAPHGIP-UHFFFAOYSA-N
SMILES: OB(O)C1=CN=C(Br)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)