CAS RN 22349-03-7
IUPAC Name:
nonadecylcyclohexane
Synonyms:
nonadecylcylohexane
Molecular Weight (g/mol):
350.68
Molecular Formula:
C25H50
InChi Key:
YVTPTBVYGTYTKP-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCCC1CCCCC1
1
–
1
of
1
results
1
Nonadecylcyclohexane 98.0+%, TCI America™
CAS: 22349-03-7 Molecular Formula: C25H50 Molecular Weight (g/mol): 350.68 MDL Number: MFCD00058954 InChI Key: YVTPTBVYGTYTKP-UHFFFAOYSA-N Synonym: 1-Cyclohexylnonadecane PubChem CID: 89671 IUPAC Name: nonadecylcyclohexane SMILES: CCCCCCCCCCCCCCCCCCCC1CCCCC1