accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22353-40-8

IUPAC Name: 2,3-dichloro-5-nitropyridine
Synonyms: 2,3-dichloro-5-nitropyridine
Molecular Weight (g/mol): 192.98
Molecular Formula: C5H2Cl2N2O2
InChi Key: XLPDVAVQKGDHNO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=C(Cl)N=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results