accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 223671-15-6

IUPAC Name: 7-bromo-1,2-dihydroisoquinolin-1-one
Synonyms: 7-bromo-2H-isoquinolin-1-one
Molecular Weight (g/mol): 224.06
Molecular Formula: C9H6BrNO
InChi Key: DSOKREQUHLPVFR-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C=CNC(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results