CAS RN 2237-30-1
3-Aminobenzonitrile, 99%
CAS: 2237-30-1 Molecular Formula: C7H6N2 Molecular Weight (g/mol): 118.139 MDL Number: MFCD00007756 InChI Key: NJXPYZHXZZCTNI-UHFFFAOYSA-N Synonym: 3-cyanoaniline,m-aminobenzonitrile,m-cyanoaniline,3-amino-benzonitrile,3-aminobenzonitril,benzonitrile, m-amino,m-anthranilonitrile,benzonitrile, 3-amino,unii-15v3cw939x,3-amino benzonitrile PubChem CID: 16702 IUPAC Name: 3-aminobenzonitrile SMILES: C1=CC(=CC(=C1)N)C#N
3-Aminobenzonitrile, 99%
CAS: 2237-30-1 Molecular Formula: C7H6N2 Molecular Weight (g/mol): 118.14 MDL Number: MFCD00007756 InChI Key: NJXPYZHXZZCTNI-UHFFFAOYSA-N Synonym: 3-cyanoaniline,m-aminobenzonitrile,m-cyanoaniline,3-amino-benzonitrile,3-aminobenzonitril,benzonitrile, m-amino,m-anthranilonitrile,benzonitrile, 3-amino,unii-15v3cw939x,3-amino benzonitrile PubChem CID: 16702 IUPAC Name: 3-aminobenzonitrile SMILES: C1=CC(=CC(=C1)N)C#N
3-Aminobenzonitrile 98.0+%, TCI America™
CAS: 2237-30-1 Molecular Formula: C7H6N2 Molecular Weight (g/mol): 118.139 MDL Number: MFCD00007756 InChI Key: NJXPYZHXZZCTNI-UHFFFAOYSA-N Synonym: 3-cyanoaniline,m-aminobenzonitrile,m-cyanoaniline,3-amino-benzonitrile,3-aminobenzonitril,benzonitrile, m-amino,m-anthranilonitrile,benzonitrile, 3-amino,unii-15v3cw939x,3-amino benzonitrile PubChem CID: 16702 IUPAC Name: 3-aminobenzonitrile SMILES: C1=CC(=CC(=C1)N)C#N
3-Aminobenzonitrile 98.0+%, TCI America™
CAS: 2237-30-1 Molecular Formula: C7H6N2 Molecular Weight (g/mol): 118.139 MDL Number: MFCD00007756 InChI Key: NJXPYZHXZZCTNI-UHFFFAOYSA-N Synonym: 3-cyanoaniline,m-aminobenzonitrile,m-cyanoaniline,3-amino-benzonitrile,3-aminobenzonitril,benzonitrile, m-amino,m-anthranilonitrile,benzonitrile, 3-amino,unii-15v3cw939x,3-amino benzonitrile PubChem CID: 16702 IUPAC Name: 3-aminobenzonitrile SMILES: C1=CC(=CC(=C1)N)C#N
Sigma Aldrich 3-Aminobenzonitrile
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.