CAS RN 2238-07-5
IUPAC Name:
2-{[(oxiran-2-yl)methoxy]methyl}oxirane
Synonyms:
diglycidyl ether
Molecular Weight (g/mol):
130.14
Molecular Formula:
C6H10O3
InChi Key:
GYZLOYUZLJXAJU-UHFFFAOYNA-N
SMILES:
C(OCC1CO1)C1CO1
1
–
1
of
1
results
1
Promotions Available
Diglycidyl ether, 95%
Diglycidyl ether, 95%
CAS: 2238-07-5 Molecular Formula: C6H10O3 Molecular Weight (g/mol): 130.14 MDL Number: MFCD00080479 InChI Key: GYZLOYUZLJXAJU-UHFFFAOYNA-N IUPAC Name: 2-{[(oxiran-2-yl)methoxy]methyl}oxirane SMILES: C(OCC1CO1)C1CO1