accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO

CAS RN 22395-22-8

IUPAC Name: 7-methoxy-2-phenyl-4H-chromen-4-one
Synonyms: 7-methoxy-2-phenylchromen-4-one
Molecular Weight (g/mol): 252.27
Molecular Formula: C16H12O3
InChi Key: QKNDCRMJDZLFEG-UHFFFAOYSA-N
SMILES: COC1=CC=C2C(=O)C=C(OC2=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results