accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH

CAS RN 22398-09-0

IUPAC Name: 4-propylpiperidine
Synonyms: 4-propyl-piperidine
Molecular Weight (g/mol): 127.23
Molecular Formula: C8H17N
InChi Key: RQGBFVLTFYRYKB-UHFFFAOYSA-N
SMILES: CCCC1CCNCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results