CAS RN 2240-27-9
CAS RN 2240-27-9
IUPAC Name:
(2R,3S,4R,5R)-5,6-dihydroxy-2,3,4-tris({[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy})hexanal
Synonyms:
(2R,3S,4R,5R)-5,6-dihydroxy-2,3,4-tris({[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy})hexanal
Molecular Weight (g/mol):
666.58
Molecular Formula:
C24H42O21
InChi Key:
MUIAGSYGABVSAA-XHFFWFORSA-N
SMILES:
OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H](O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@@H](O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)C=O
1
–
2
of
2
results
1
D-(+)-Cellopentaose, 95%, Thermo Scientific Chemicals
CAS: 2240-27-9 MDL Number: MFCD09750881 Synonym: Cellopentaose
2
Cellopentaose 95.0+%, TCI America™
CAS: 2240-27-9 Molecular Formula: C30H52O26 MDL Number: MFCD00151164 Synonym: Glc beta(1-4)Glc beta(1-4)Glc beta(1-4)Glc beta(1-4)Glc