accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COPOOCH3

CAS RN 2240-41-7

IUPAC Name: dimethyl phenylphosphonate
Synonyms: O,O-dimethylphenylphosphonate
Molecular Weight (g/mol): 186.15
Molecular Formula: C8H11O3P
InChi Key: OXDOANYFRLHSML-UHFFFAOYSA-N
SMILES: COP(=O)(OC)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results