accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3NH

CAS RN 22401-74-7

IUPAC Name: 3-tert-butyl-9H-carbazole
Synonyms: 3-tert-butyl-9H-carbazole
Molecular Weight (g/mol): 223.32
Molecular Formula: C16H17N
InChi Key: TYXSZNGDCCGIBO-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC2=C(NC3=C2C=CC=C3)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results