accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 224047-41-0

IUPAC Name: 4-(4-chlorophenyl)-2-phenyl-3-(1H-1,2,4-triazol-1-yl)butan-2-ol
Synonyms: 4-(4-chlorophenyl)-2-phenyl-3-(1,2,4-triazol-1-yl)butan-2-ol
Molecular Weight (g/mol): 327.81
Molecular Formula: C18H18ClN3O
InChi Key: YULDTPKHZNKFEY-UHFFFAOYNA-N
SMILES: CC(O)(C(CC1=CC=C(Cl)C=C1)N1C=NC=N1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results